DL-3-Methyl-2-butanol, 98%
DL-3-Methyl-2-butanol, 98%
DL-3-Methyl-2-butanol, 98%
DL-3-Methyl-2-butanol, 98%
Thermo Scientific Chemicals

DL-3-Methyl-2-butanol, 98%

CAS: 598-75-4 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 149930250
Price (USD)
32.65
Online Exclusive
35.60
Save 2.95 (8%)
Each
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
32.65
Online Exclusive
35.60
Save 2.95 (8%)
Each
Add to cart
Chemical Identifiers
CAS598-75-4
IUPAC Name3-methylbutan-2-ol
Molecular FormulaC5H12O
InChI KeyMXLMTQWGSQIYOW-UHFFFAOYNA-N
SMILESCC(C)C(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4080 to 1.4100 (20°C, 589 nm)
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection