Vinylcyclopentane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Vinylcyclopentane, 97%, Thermo Scientific Chemicals
 
Have Questions?
Quantity:
1 g
Packaging:
Glass bottle
Catalog number 150050010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3742-34-5
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless
Appearance (Form)Liquid
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only