Thermo Scientific Chemicals

trans-beta-Methylstyrene, 97%, stabilized, Thermo Scientific Chemicals

Catalog number: 150150500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

trans-beta-Methylstyrene, 97%, stabilized, Thermo Scientific Chemicals

Catalog number: 150150500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 150150500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
873-66-5
IUPAC Name
[(1E)-prop-1-en-1-yl]benzene
Molecular Formula
C9H10
InChI Key
QROGIFZRVHSFLM-QHHAFSJGSA-N
SMILES
CC=CC1=CC=CC=C1
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=96.0 %
Refractive index
1.5480 to 1.5500 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text