Thermo Scientific Chemicals

4-Chlorobenzoyl chloride, 99+%, Thermo Scientific Chemicals

Catalog number: 150191000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

4-Chlorobenzoyl chloride, 99+%, Thermo Scientific Chemicals

Catalog number: 150191000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 150191000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
122-01-0
IUPAC Name
4-chlorobenzoyl chloride
Molecular Formula
C7H4Cl2O
InChI Key
RKIDDEGICSMIJA-UHFFFAOYSA-N
SMILES
ClC(=O)C1=CC=C(Cl)C=C1
Refractive index
1.5770 to 1.5790 (20°C, 589 nm)
Appearance (Color)
Clear colorless to slightly colored
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Titration RCOCl
>=98.50 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text