3,4-Dichlorobenzoyl chloride, 97%, Thermo Scientific Chemicals
3,4-Dichlorobenzoyl chloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,4-Dichlorobenzoyl chloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass Bottle
Catalog number 150231000
Price (USD)/ Each
146.00
-
Add to cart
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
146.00
Add to cart
3,4-Dichlorobenzoyl chloride, 97%, Thermo Scientific Chemicals
Catalog number150231000
Price (USD)/ Each
146.00
-
Add to cart
Chemical Identifiers
IUPAC Name3,4-dichlorobenzoyl chloride
Molecular FormulaC7H3Cl3O
InChI KeyVTXNOVCTHUBABW-UHFFFAOYSA-N
SMILESClC(=O)C1=CC=C(Cl)C(Cl)=C1
Molecular Weight (g/mol)209.45
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Low melting crystalline solid
Infrared spectrumConforms
Titration RCOCl>=96.0 %
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only