Search by lot number or partial lot number
Closure Material | LDPE |
Diameter (Metric) | 17.5 mm |
Max. RCF | 6000 x g |
Product Type | Round Bottom Tube |
Certifications/Compliance | Mouse Embryo Assay (1-cell stage MEA) tested. Produced under ISO 13485 with full lot traceability. USP Class VI tested. Non-pyrogenic and sterile (SAL 10-6). |
Closure Type | Dual Position Vent/Close Cap |
Length (Metric) | 95 mm |
Material | Polystyrene |
No. per Pack | 10 |
No. per Case | 500 |
Sterility | Sterile |
Capacity (Metric) | 14 mL |
Shape | Round |
Closure Included | Yes |
Unit Size | Case of 500 |
Catalog Number | Specifications | Unit Size | No. per Case | No. per Pack | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
150268 | Case of 500 | 500 | 10 | 292.65 Online price 307.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|