1x1 image pixel for data collection
2-Methyl-2-propen-1-ol, 98%
2-Methyl-2-propen-1-ol, 98%
2-Methyl-2-propen-1-ol, 98%
2-Methyl-2-propen-1-ol, 98%
Thermo Scientific Chemicals

2-Methyl-2-propen-1-ol, 98%

CAS: 513-42-8 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
100 mL
Catalog number 150290100
Price (USD)
-
Quantity:
10 mL
Request bulk or custom format
Chemical Identifiers
CAS513-42-8
IUPAC Name2-methylprop-2-en-1-ol
Molecular FormulaC4H8O
InChI KeyBYDRTKVGBRTTIT-UHFFFAOYSA-N
SMILESCC(=C)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4250 to 1.4270 (20°C, 589 nm)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link