Thermo Scientific Chemicals

1,1,1-Trichloro-2-methyl-2-propanol hemihydrate, 98%, Thermo Scientific Chemicals

Catalog number: 150301000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

1,1,1-Trichloro-2-methyl-2-propanol hemihydrate, 98%, Thermo Scientific Chemicals

Catalog number: 150301000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 150301000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6001-64-5
IUPAC Name
bis(1,1,1-trichloro-2-methylpropan-2-ol) hydrate
Molecular Formula
C8H16Cl6O3
InChI Key
WRWLCXJYIMRJIN-UHFFFAOYSA-N
SMILES
O.CC(C)(O)C(Cl)(Cl)Cl.CC(C)(O)C(Cl)(Cl)Cl
Appearance (Form)
Crystalline powder or crystals and/or chunks
Infrared spectrum
Conforms
Titration Argentometric
97.5 to 102.5 % (ex Cl)
Appearance (Color)
White to off-white
Water
4.5 to 5.5 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text