Thermo Scientific™

Pierce™ 8-Well Polystyrene Strip Plates, Corner Notch

Catalog number: 15031
Have Questions?
Thermo Scientific™

Pierce™ 8-Well Polystyrene Strip Plates, Corner Notch

Catalog number: 15031
Have Questions?
Catalog Number
15031
Unit Size
100 plates
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
15031100 plates
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce 8-Well Strip Plates are optically clear, polystyrene plates that have a standard 96-well microplate footprint but also allow individual 8-well strips (i.e., plate columns) to be removed from the rigid frame.

Features of 8-Well Strip Plates:

• Special optically clear polystyrene formulation
• High antibody-binding surface improves the overall performance in immunoassay applications
• Plates are certified for performance, having consistently low coefficients of variation among wells
• Eight-well strips fit snugly in the plate frame, virtually eliminating strip loss while in use—even if turned upside down
• Strip guides help to avoid misorientation of the strip when returned to the holder
• Strips are gently removed from the frame with a strip-well ejector
• Wells can be easily separated from the strips with a gentle twist, allowing you to work with individual wells or strips of less than eight wells

The format provides convenience and quality-assurance when performing partial-plate assays because unused wells can be removed to prevent confusion and cross-contamination.

Plate Properties:
• Antibody binding capacity: High, 400ng IgG/cm2 (1 cm2 = 100 μL volume)
• Binding interaction: hydrophobic-ionic
• Well-to-well coefficient of variation: CV ≤3%
• Well shape (bottom): Flat
• Maximum well volume: 360 μL
• Packaging: 5 plates per sealed tray
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Binding Property
Adsorption, Hydrophilic
Detection Method
Colorimetric
Packaging
100 Plate
Product Line
Pierce
Color
Clear
Product Type
Well Plate
Material
Polystyrene
Volume (Metric) Well
360 μL
No. of Wells
96
Surface Treatment
Polystyrene
Well Shape
Flat Bottom (F-well)
Plate Blocking
None

Contents & Storage

Contents: 100 plates, 5 plates per sealed tray

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text