Search Thermo Fisher Scientific
Catalog Number | Culture Area | Surface Treatment | Gridded | Vent |
---|---|---|---|---|
150318 | 8.8 sq. cm | Nunclon Delta | Not Gridded | Lid without airvent |
153066 | 8.8 sq. cm | Nunclon Delta | Not Gridded | Lid with airvent |
174926 | 8.8 sq. cm | Nunclon Delta | Gridded | Lid with airvent |
171099 | 8.8 sq. cm | Non-treated | Not Gridded | Lid with airvent |
174888 | 20.8 sq. cm | Cell Culture | Not Gridded | Lid with airvent |
150326 | 21.5 sq. cm | Nunclon Delta | Not Gridded | Lid without airvent |
150288 | 21.5 sq. cm | Nunclon Delta | Not Gridded | Lid with airvent |
169558 | 21.5 sq. cm | Nunclon Delta | Gridded | Lid with airvent |
150350 | 56.7 sq. cm | Nunclon Delta | Not Gridded | Lid with airvent |
168381 | 145 sq. cm | Nunclon Delta | Not Gridded | Lid with airvent |
172931 | 56.7 sq. cm | Nunclon Delta | Not Gridded | Lid with airvent |
Experience optimal performance across a broad range of sizes and styles to suit a variety of experimental needs. Thermo Scientific Nunc Cell Culture Dishes receive the Nunclon Delta surface treatment, a proprietary surface modification that promotes maximum adhesion for a broad range of cell types. The Petri dishes are made from high quality and optically clear polystyrene.
Nunc culture dishes, available in a variety of sizes and designs, are made from high quality optically clear polystyrene or Permanox plastic.
Ordering information
Cat. Nos. 174926 and 169558 feature 2mm grids
Cat. Nos. 171099 and 150340 are for suspension culture and not Nunclon-treated
Includes a certificate that assures user of biological performance as well as traceability.
Share catalog number, name or link