(1S)-(-)-β-pinène, 98 %, Thermo Scientific Chemicals
(1S)-(-)-β-pinène, 98 %, Thermo Scientific Chemicals
(1S)-(-)-β-pinène, 98 %, Thermo Scientific Chemicals
(1S)-(-)-β-pinène, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(1S)-(-)-β-pinène, 98 %, Thermo Scientific Chemicals

CAS: 18172-67-3 | C10H16 | 136.24 g/mol
Have Questions?
Modifier l'affichagebuttonViewtableView
Quantité:
250 mL
1 L
Conditionnement:
Flacon en verre
Référence 150322500
Prix (EUR)
27,37
Offre exceptionnelle en ligne
32,20
Économisez 4,83 (15%)
Each
-
Ajouter au panier
Quantité:
250 mL
Conditionnement:
Flacon en verre
Recurring order eligible. Learn more »
Grand volume ou format personnalisé
Prix (EUR)
27,37
Offre exceptionnelle en ligne
32,20
Économisez 4,83 (15%)
Each
Ajouter au panier
Identifiants chimiques
CAS18172-67-3
IUPAC Name(1S,5S)-6,6-dimethyl-2-methylidenebicyclo[3.1.1]heptane
Molecular FormulaC10H16
InChI KeyWTARULDDTDQWMU-IUCAKERBSA-N
SMILESCC1(C)[C@@H]2C[C@H]1C(=C)CC2
Voir plus
Specific optical rotation-17° to -22° (25°C, 589 nm) (neat)
Specific gravity(25°C) 0.860 to 0.880
Appearance (Color)Clear colorless to light yellow
Refractive index1.4770 to 1.4810 (20°C, 589 nm)
Infrared spectrumConforms
Voir plus
Ask our AI about this Product

Ce produit de la marque Thermo Scientific Chemicals faisait à l’origine partie de la gamme de produits Acros Organics. Certains documents et informations sur l’étiquette peuvent faire référence à la marque héritée. Le code d’article / produit Acros Organics d’origine ou la référence SKU n’a pas changé dans le cadre de la transition de la marque vers Thermo Scientific Chemicals.

RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documentation et téléchargements

Certificats

Numéro de lotCertificate TypeDateCatalog Number(s)
A0470362Certificate of Analysis17 mai 2025150320010, 150322500
A0465161Certificate of Analysis06 août 2024150320010, 150322500
A0440992Certificate of Analysis15 avr. 2024150320010, 150322500
2107919Certificate of Analysis12 déc. 2022150320010, 150322500
4109A44Certificate of Analysis12 déc. 2022150320010, 150322500
5 résultats affichés, recherchez un certificat spécifique ci-dessus

Safety Data Sheets

Sécurité et manipulation

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Flammable liquids
Category 3
Skin Corrosion/Irritation
Category 2
Skin Sensitization
Category 1
Aspiration Toxicity
Category 1
Acute aquatic toxicity
Category 1
Chronic aquatic toxicity
Category 1
Label Elements
Signal Word

Danger

Hazard Statements

H304 - May be fatal if swallowed and enters airways

H315 - Causes skin irritation

H317 - May cause an allergic skin reaction

H410 - Very toxic to aquatic life with long lasting effects

Physical Hazards

H226 - Flammable liquid and vapor

Precautionary Statements

P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

P264 - Wash face, hands and any exposed skin thoroughly after handling

P280 - Wear protective gloves/protective clothing

P301 + P310 - IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician

P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

P331 - Do NOT induce vomiting

P332 + P313 - If skin irritation occurs: Get medical advice/attention

P333 + P313 - If skin irritation or rash occurs: Get medical advice/attention

Partager la référence catalogue, le nom ou le lien.

1x1 image pixel for data collection