Thymol, 99%, Thermo Scientific Chemicals
Save 25% on this product with promo code P5488313
Thymol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Thymol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Plastic bottle
Catalog number 150330025
Price (USD)/ Each
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name5-methyl-2-(propan-2-yl)phenol
Molecular FormulaC10H14O
InChI KeyMGSRCZKZVOBKFT-UHFFFAOYSA-N
SMILESCC(C)C1=CC=C(C)C=C1O
Molecular Weight (g/mol)150.22
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point48°C to 52°C
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only