Thermo Scientific Chemicals

Thymol, 99%, Thermo Scientific Chemicals

Catalog number: 150331000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Thymol, 99%, Thermo Scientific Chemicals

Catalog number: 150331000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 150331000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
89-83-8
IUPAC Name
5-methyl-2-(propan-2-yl)phenol
Molecular Formula
C10H14O
InChI Key
MGSRCZKZVOBKFT-UHFFFAOYSA-N
SMILES
CC(C)C1=CC=C(C)C=C1O
Appearance (Color)
White to off-white
Appearance (Form)
Crystalline powder or crystals
Infrared spectrum
Conforms
Melting point
48°C to 52°C
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text