Nunc™ IVF Dishes
Save 51% on plasticware essentials and stock your lab to culture with confidence. View promotion details >
Nunc™ IVF Dishes
Nunc™ IVF Dishes
Nunc™ IVF Dishes
Nunc™ IVF Dishes
Thermo Scientific™

Nunc™ IVF Dishes

Carry out IVF applications using high-quality, reliable Thermo Scientific™ Nunc™ IVF Dishes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Height (Metric)Culture Area
15036088 mm15 mm56.7 cm2
15025535 mm11 mm8.8 cm2
15027054 mm14 mm21.5 cm2
Catalog number 150360
Price (USD)
461.65
Online Exclusive
475.00
Save 13.35 (3%)
Case of 150
-
Add to cart
Diameter (Metric):
88 mm
Height (Metric):
15 mm
Culture Area:
56.7 cm2
Price (USD)
461.65
Online Exclusive
475.00
Save 13.35 (3%)
Case of 150
Add to cart
Ask our AI about this Product

Carry out IVF applications using high-quality, reliable Thermo Scientific™ Nunc™ IVF Dishes.

Product features for reliable results:

  • Facilitate gas exchange with vented lid
  • Available in 35mm, 60mm and 90mm sizes
  • Certificates of conformity available for every lot

Rigorously tested, certified and validated for IVF:

  • CE-marked (Notified Body 2460), FDA-listed
  • Produced under ISO 13485 with full lot traceability
  • Sterile (SAL 10-6)
  • Non-pyrogenic (LAL tested)
  • USP Class VI polystyrene
  • Lot-release tested by 1-cell stage Mouse Embryo Assay (MEA) and Human Sperm Survival Assay (HSSA)

Warranty: 90 days

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceCE-marked (Notified Body 2460), FDA-listed, MEA-tested, HSSA-tested
DescriptionIVF Dish, 90mm, Lid w/Airvent
No. per Case150
No. per Pack10
Culture Area56.7 cm2
Diameter (Metric)88 mm
Height (Metric)15 mm
MaterialPolystyrene (USP Class VI)
SterilitySterile
Surface TreatmentNon-treated
Unit SizeCase of 150
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection