Thermo Scientific Chemicals

2-Methylbenzothiazole, 99%, Thermo Scientific Chemicals

Catalog number: 150380250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Methylbenzothiazole, 99%, Thermo Scientific Chemicals

Catalog number: 150380250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 150380250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
120-75-2
IUPAC Name
2-methyl-1,3-benzothiazole
Molecular Formula
C8H7NS
InChI Key
DXYYSGDWQCSKKO-UHFFFAOYSA-N
SMILES
CC1=NC2=CC=CC=C2S1
Appearance (Color)
Clear colorless to yellow to orange-brown
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=98.5 %
Refractive index
1.6150 to 1.6180 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text