2-Methylbenzothiazole, 99%, Thermo Scientific Chemicals
2-Methylbenzothiazole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methylbenzothiazole, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number 150381000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS120-75-2
IUPAC Name2-methyl-1,3-benzothiazole
Molecular FormulaC8H7NS
InChI KeyDXYYSGDWQCSKKO-UHFFFAOYSA-N
SMILESCC1=NC2=CC=CC=C2S1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to orange-brown
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.6150 to 1.6180 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only