Thermo Scientific Chemicals

2,4-Dichloroaniline, 98%, Thermo Scientific Chemicals

Catalog number: 150390250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2,4-Dichloroaniline, 98%, Thermo Scientific Chemicals

Catalog number: 150390250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 150390250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
554-00-7
IUPAC Name
2,4-dichloroaniline
Molecular Formula
C6H5Cl2N
InChI Key
KQCMTOWTPBNWDB-UHFFFAOYSA-N
SMILES
NC1=CC=C(Cl)C=C1Cl
Appearance (Color)
Beige to light grey to light brown
Appearance (Form)
Powder or crystals or flakes and/or chunks
Appearance
May darken on storage
Infrared spectrum
Conforms
Melting point
59°C to 63°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text