Versene Solution
Versene Solution
Gibco™

Versene Solution

Versene is an EDTA solution for use as a gentle non-enzymatic cell dissociation reagent. Gibco Versene Solution (0.48 mM) isRead more
Have Questions?
Catalog NumberQuantity
15040066100 mL
Catalog number 15040066
Price (EUR)
22,59
Each
-
Add to cart
Quantity:
100 mL
Price (EUR)
22,59
Each
Add to cart
Ask our AI about this Product
Versene is an EDTA solution for use as a gentle non-enzymatic cell dissociation reagent. Gibco Versene Solution (0.48 mM) is formulated as 0.2 g EDTA(Na4) per liter of Phosphate Buffered Saline (PBS).

Product Use
For Research Use Only. Not for use in diagnostic procedures.

cGMP Manufacturing and Quality System
For supply chain continuity, we manufacture Gibco Versene Solution at two separate facilities located in Grand Island, NY and Scotland, UK. Both sites are compliant with cGMP manufacturing requirements and are certified to the ISO 13485 standard.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ChelatorsEDTA
Manufacturing QualitycGMP for medical devices, 21 CFR Part 820 and ISO 13485
Quantity100 mL
Shelf Life18 Months
Shipping ConditionRoom Temperature
ClassificationAnimal Origin-free
Product TypeCell Culture Dissociation Reagent
SterilitySterile-filtered
With AdditivesEDTA
Unit SizeEach
Contents & Storage
Storage conditions: 2 to 8°C
Shipping conditions: Ambient
Shelf life: 18 months from date of manufacture
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link