Thermo Scientific™

Pierce™ 96-Well Polystyrene Plates, Corner Notch

Catalog number: 15041
Have Questions?
Thermo Scientific™

Pierce™ 96-Well Polystyrene Plates, Corner Notch

Catalog number: 15041
Have Questions?
Catalog Number
15041
Unit Size
100 plates
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
15041100 plates
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce Clear 96-Well Microplates are made of specially formulated polystyrene and treated to produce high-capacity antibody-binding characteristics. These microplates have a corner notch for use in standard plate readers.

Features of Clear 96-Well Microplates:

• Clear plates for colorimetric ELISA and other assays
• Specially formulated optically clear polystyrene and treated to provide protein coating capacity
• High antibody-binding surface improves the overall performance in immunoassay applications
• Plates are certified for performance, having consistently low coefficients of variation among wells

Plate Properties:
• Antibody binding capacity: High, 400ng IgG/cm2 (1 cm2 = 100 μL volume)
• Binding interaction: hydrophobic-ionic
• Well-to-well coefficient of variation: CV ≤3%
• Well shape (bottom): Flat
• Maximum well volume: 360 μL

Related Products
Pierce™ 96-Well Polystyrene Plates, White Opaque
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Binding Property
Adsorption, Hydrophilic
Description
Pierce 96-Well Polystyrene Plates, Corner Notch
Detection Method
Colorimetric
Packaging
100 Plate
Product Line
Pierce
Color
Clear
Product Type
Well Plate
Material
Polystyrene
Volume (Metric) Well
360 μL
No. of Wells
96
Surface Treatment
Polystyrene
Well Shape
Flat Bottom (F-well)
Plate Blocking
None

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text