Thermo Scientific Chemicals

4-Bromobenzyl bromide, 98%, Thermo Scientific Chemicals

Catalog number: 150410250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Bromobenzyl bromide, 98%, Thermo Scientific Chemicals

Catalog number: 150410250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 150410250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
589-15-1
IUPAC Name
1-bromo-4-(bromomethyl)benzene
Molecular Formula
C7H6Br2
InChI Key
YLRBJYMANQKEAW-UHFFFAOYSA-N
SMILES
BrCC1=CC=C(Br)C=C1
Appearance (Form)
Powder or crystals or needles and/or chunks
Melting point
60°C to 64°C
Appearance (Color)
White to orange to tan
GC
>=97.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text