4-Nitrobenzoyl chloride, 98%, Thermo Scientific Chemicals
4-Nitrobenzoyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Nitrobenzoyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
250 g
Packaging:
Glass bottle
Catalog number 150435000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS122-04-3
IUPAC Name4-nitrobenzoyl chloride
Molecular FormulaC7H4ClNO3
InChI KeySKDHHIUENRGTHK-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(C=C1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Appearance (Form)Crystals or flakes
Infrared spectrumConforms
Melting point70°C to 74°C
Titration RCOCl>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only