4-Chlorobenzaldehyde, 98%
4-Chlorobenzaldehyde, 98%
4-Chlorobenzaldehyde, 98%
4-Chlorobenzaldehyde, 98%
Thermo Scientific Chemicals

4-Chlorobenzaldehyde, 98%

CAS: 104-88-1 | C7H5ClO | 140.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 150442500
Price (USD)
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS104-88-1
IUPAC Name4-chlorobenzaldehyde
Molecular FormulaC7H5ClO
InChI KeyAVPYQKSLYISFPO-UHFFFAOYSA-N
SMILESClC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Color)White to light yellow
Infrared spectrumConforms
Free acid=<1.5 % (as 4-Chlorobenzoic acid)
Appearance (Form)Crystals or crystalline or fused solid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link