Thermo Scientific Chemicals

Ethyl propionate, 99+%, Thermo Scientific Chemicals

Catalog number: 150500250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl propionate, 99+%, Thermo Scientific Chemicals

Catalog number: 150500250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 150500250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
105-37-3
IUPAC Name
ethyl propanoate
Molecular Formula
C5H10O2
InChI Key
FKRCODPIKNYEAC-UHFFFAOYSA-N
SMILES
CCOC(=O)CC
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.3830 to 1.3870 (20°C, 589 nm)
Acid value
=<1 mg KOH/g
Appearance (Color)
Clear colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text