Ethyl Caprylate, 99+%, Thermo Scientific Chemicals
Ethyl Caprylate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl Caprylate, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 150510050
Price (USD)/ Each
16.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
19.70 
Save 3.05 (15%)
-
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
16.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
19.70 
Save 3.05 (15%)
Add to cart
Ethyl Caprylate, 99+%, Thermo Scientific Chemicals
Catalog number150510050
Price (USD)/ Each
16.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
19.70 
Save 3.05 (15%)
-
Add to cart
Chemical Identifiers
CAS106-32-1
IUPAC Nameethyl octanoate
Molecular FormulaC10H20O2
InChI KeyYYZUSRORWSJGET-UHFFFAOYSA-N
SMILESCCCCCCCC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
GC>=99.0 %
Water=<0.3 % (K.F.)
Specific gravity(20°C) 0.866 to 0.87
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only