Thermo Scientific Chemicals

Ethyl Caprylate, 99+%, Thermo Scientific Chemicals

Catalog number: 150515000
500 mL, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl Caprylate, 99+%, Thermo Scientific Chemicals

Catalog number: 150515000
500 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 150515000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
106-32-1
IUPAC Name
ethyl octanoate
Molecular Formula
C10H20O2
InChI Key
YYZUSRORWSJGET-UHFFFAOYSA-N
SMILES
CCCCCCCC(=O)OCC
GC
>=99.0 %
Water
=<0.3 % (K.F.)
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
Specific gravity
(20°C) 0.866 to 0.87

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text