Thermo Scientific Chemicals

Ethyl cinnamate, 98% trans, Thermo Scientific Chemicals

Catalog number: 150535000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl cinnamate, 98% trans, Thermo Scientific Chemicals

Catalog number: 150535000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 150535000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
103-36-6
IUPAC Name
ethyl (2E)-3-phenylprop-2-enoate
Molecular Formula
C11H12O2
InChI Key
KBEBGUQPQBELIU-CMDGGOBGSA-N
SMILES
CCOC(=O)\C=C\C1=CC=CC=C1
Specific gravity
(25/25°C) 1.045 to 1.048
Acid value
=<1 mg KOH/g
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text