This product ships via ground from US northeast.
D(+)-Mannose, 99+%
Thermo Scientific Chemicals

D(+)-Mannose, 99+%

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
25 g
100 g
1 kg
Packaging:
View all
Plastic drum
Catalog number 150600025
Price (USD)/ Each
1,582.65
Online exclusive
1862.00 
Save 279.35 (15%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
1,582.65
Online exclusive
1862.00 
Save 279.35 (15%)
Add to cart
D(+)-Mannose, 99+%
Catalog number150600025
Price (USD)/ Each
1,582.65
Online exclusive
1862.00 
Save 279.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name6-(hydroxymethyl)oxane-2,3,4,5-tetrol
Molecular FormulaC6H12O6
InChI KeyWQZGKKKJIJFFOK-UHFFFAOYNA-N
SMILESOCC1OC(O)C(O)C(O)C1O
Molecular Weight (g/mol)180.16
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance of solution(10 % in water) Clear
Bacterial endotoxins(LAL-test) 0.5 E.U. in 1 ml of a 0.1% soln.
Appearance (Form)Powder or crystals
Infrared spectrumConforms
HPLC>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only