Informational:Informational:This product ships via ground from US northeast.
Thermo Scientific Chemicals

Histamine dihydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 150620050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Histamine dihydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 150620050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 150620050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56-92-8
IUPAC Name
dihydrogen 2-(1H-imidazol-5-yl)ethan-1-amine dichloride
Molecular Formula
C5H11Cl2N3
InChI Key
PPZMYIBUHIPZOS-UHFFFAOYSA-N
SMILES
[H+].[H+].[Cl-].[Cl-].NCCC1=CN=CN1
Loss on drying
=<0.5 % (105°C)
Sulfated ash
=<0.1 %
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text