Histamine dihydrochloride, 99%, Thermo Scientific Chemicals
This product ships via ground from US northeast.
Histamine dihydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Histamine dihydrochloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 150625000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS56-92-8
IUPAC Namedihydrogen 2-(1H-imidazol-5-yl)ethan-1-amine dichloride
Molecular FormulaC5H11Cl2N3
InChI KeyPPZMYIBUHIPZOS-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].NCCC1=CN=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfated ash=<0.1 %
Loss on drying=<0.5 % (105°C)
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only