Thermo Scientific Chemicals

1-Octanol, 99%, pure, Thermo Scientific Chemicals

Catalog number: 150630250
25 L, Metal Drum, Each
Thermo Scientific Chemicals

1-Octanol, 99%, pure, Thermo Scientific Chemicals

Catalog number: 150630250
25 L, Metal Drum, Each
Quantity
Packaging
Catalog number: 150630250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-87-5
IUPAC Name
octan-1-ol
Molecular Formula
C8H18O
InChI Key
KBPLFHHGFOOTCA-UHFFFAOYSA-N
SMILES
CCCCCCCCO
GC
>=98.5 %
Refractive index
1.4280 to 1.4310 (20°C, 589 nm)
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text