Thermo Scientific Chemicals

2-Methoxy-4-nitroaniline, 98%, Thermo Scientific Chemicals

Catalog number: 150641000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

2-Methoxy-4-nitroaniline, 98%, Thermo Scientific Chemicals

Catalog number: 150641000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 150641000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
97-52-9
IUPAC Name
2-methoxy-4-nitroaniline
Molecular Formula
C7H8N2O3
InChI Key
GVBHRNIWBGTNQA-UHFFFAOYSA-N
SMILES
COC1=CC(=CC=C1N)[N+]([O-])=O
Appearance (Color)
Yellow to orange to brown
Appearance (Form)
Powder or crystalline powder or crystals
Infrared spectrum
Conforms
Melting point
138°C to 142°C
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text