Thermo Scientific Chemicals

4-Butylaniline, 97%, Thermo Scientific Chemicals

Catalog number: 150651000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Butylaniline, 97%, Thermo Scientific Chemicals

Catalog number: 150651000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 150651000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
104-13-2
IUPAC Name
4-butylaniline
Molecular Formula
C10H15N
InChI Key
OGIQUQKNJJTLSZ-UHFFFAOYSA-N
SMILES
CCCCC1=CC=C(N)C=C1
Appearance (Color)
Clear yellow to light brown
Appearance (Form)
Liquid
GC
>=96.0 %
Infrared spectrum
Conforms
Refractive index
1.5340 to 1.5360 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text