Thermo Scientific Chemicals

Isoamyl acetate, 99+%, pure, Thermo Scientific Chemicals

Catalog number: 150660025
2.5 L, Glass bottle, Each
Thermo Scientific Chemicals

Isoamyl acetate, 99+%, pure, Thermo Scientific Chemicals

Catalog number: 150660025
2.5 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 150660025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
123-92-2
IUPAC Name
3-methylbutyl acetate
Molecular Formula
C7H14O2
InChI Key
MLFHJEHSLIIPHL-UHFFFAOYSA-N
SMILES
CC(C)CCOC(C)=O
Infrared spectrum
Conforms
Refractive index
1.3990 to 1.4030 (20°C, 589 nm)
GC
>=98.5 % (sum of isomers)
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text