1x1 image pixel for data collection
Ethyl formate, 98+%, pure
Ethyl formate, 98+%, pure
Ethyl formate, 98+%, pure
Ethyl formate, 98+%, pure
Thermo Scientific Chemicals

Ethyl formate, 98+%, pure

CAS: 109-94-4 | C3H6O2 | 74.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 150675000
Price (USD)
57.65
Online Exclusive
64.20
Save 6.55 (10%)
Each
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
57.65
Online Exclusive
64.20
Save 6.55 (10%)
Each
Add to cart
Chemical Identifiers
CAS109-94-4
IUPAC Nameethyl formate
Molecular FormulaC3H6O2
InChI KeyWBJINCZRORDGAQ-UHFFFAOYSA-N
SMILESCCOC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.3580 to 1.3630 (20°C, 589 nm)
GC>=98.0 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link