Formaldehyde Sodium Bisulfite Addition Compound 95%, Thermo Scientific Chemicals
Formaldehyde Sodium Bisulfite Addition Compound 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Formaldehyde Sodium Bisulfite Addition Compound 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 g
5 kg
Packaging:
View all
Plastic bottle
Catalog number 150690010
Price (USD)/ Each
47.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
55.30 
Save 7.65 (14%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
47.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
55.30 
Save 7.65 (14%)
Add to cart
Formaldehyde Sodium Bisulfite Addition Compound 95%, Thermo Scientific Chemicals
Catalog number150690010
Price (USD)/ Each
47.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
55.30 
Save 7.65 (14%)
-
Add to cart
Chemical Identifiers
CAS870-72-4
IUPAC Namesodium hydroxymethanesulfonate
Molecular FormulaCH3NaO4S
InChI KeyUOULCEYHQNCFFH-UHFFFAOYSA-M
SMILES[Na+].OCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Titration>=94 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only