Thermo Scientific Chemicals

Dimethyl succinate, 99%, Thermo Scientific Chemicals

Catalog number: 150731000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Dimethyl succinate, 99%, Thermo Scientific Chemicals

Catalog number: 150731000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 150731000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
106-65-0
IUPAC Name
1,4-dimethyl butanedioate
Molecular Formula
C6H10O4
InChI Key
MUXOBHXGJLMRAB-UHFFFAOYSA-N
SMILES
COC(=O)CCC(=O)OC
Infrared spectrum
Conforms
Refractive index
1.4180 to 1.4210 (20°C, 589 nm)
Appearance (Form)
Clear liquid
Color scale
=<25 APHA
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text