Dimethyl succinate, 99%, Thermo Scientific Chemicals
Dimethyl succinate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethyl succinate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number 150735000
Price (USD)/ Each
93.65
Online exclusive
104.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
93.65
Online exclusive
104.00 
Save 10.35 (10%)
Add to cart
Dimethyl succinate, 99%, Thermo Scientific Chemicals
Catalog number150735000
Price (USD)/ Each
93.65
Online exclusive
104.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS106-65-0
IUPAC Name1,4-dimethyl butanedioate
Molecular FormulaC6H10O4
InChI KeyMUXOBHXGJLMRAB-UHFFFAOYSA-N
SMILESCOC(=O)CCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<25 APHA
Infrared spectrumConforms
Refractive index1.4180 to 1.4210 (20°C, 589 nm)
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only