Thermo Scientific Chemicals

Dicyclopentadiene, 95%, stabilized with 100-200 ppm 4-tert-Butylcatechol, Thermo Scientific Chemicals

Catalog number: 150760100
10 L, Plastic drum, Each
Thermo Scientific Chemicals

Dicyclopentadiene, 95%, stabilized with 100-200 ppm 4-tert-Butylcatechol, Thermo Scientific Chemicals

Catalog number: 150760100
10 L, Plastic drum, Each
Quantity
Packaging
Catalog number: 150760100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
77-73-6
IUPAC Name
tricyclo[5.2.1.0²,⁶]deca-3,8-diene
Molecular Formula
C10H12
InChI Key
HECLRDQVFMWTQS-UHFFFAOYNA-N
SMILES
C1C=CC2C3CC(C=C3)C12
Color scale
=<50 APHA
Infrared spectrum
Conforms
GC
>=91 % (endo-Dicyclopentadiene)
Water
=<0.03 % (K.F.)
Appearance (Form)
Clear liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text