Thermo Scientific Chemicals

Ethyl 4-aminobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: 150780025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Ethyl 4-aminobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: 150780025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 150780025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
94-09-7
IUPAC Name
ethyl 4-aminobenzoate
Molecular Formula
C9H11NO2
InChI Key
BLFLLBZGZJTVJG-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CC=C(N)C=C1
Appearance (Form)
Crystalline powder
GC
>=97.5 %
Melting point
89°C to 92°C
Infrared spectrum
Conforms
Titration with HClO4
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text