Ethyl 4-aminobenzoate, 98%
Ethyl 4-aminobenzoate, 98%
Ethyl 4-aminobenzoate, 98%
Ethyl 4-aminobenzoate, 98%
Thermo Scientific Chemicals

Ethyl 4-aminobenzoate, 98%

CAS: 94-09-7 | C9H11NO2 | 165.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 150785000
Price (USD)
167.65
Online Exclusive
186.00
Save 18.35 (10%)
Each
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
167.65
Online Exclusive
186.00
Save 18.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS94-09-7
IUPAC Nameethyl 4-aminobenzoate
Molecular FormulaC9H11NO2
InChI KeyBLFLLBZGZJTVJG-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
GC>=97.5 %
Melting point89°C to 92°C
Infrared spectrumConforms
Titration with HClO4>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection