Thermo Scientific Chemicals

Acetanilide, 99+%, Thermo Scientific Chemicals

Catalog number: 150810010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Acetanilide, 99+%, Thermo Scientific Chemicals

Catalog number: 150810010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 150810010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
103-84-4
IUPAC Name
N-phenylacetamide
Molecular Formula
C8H9NO
InChI Key
FZERHIULMFGESH-UHFFFAOYSA-N
SMILES
CC(=O)NC1=CC=CC=C1
Infrared spectrum
Conforms
GC
>=99.0 %
Melting point
111°C to 115°C
Appearance (Form)
Powder or crystals or flakes
Appearance (Color)
White to beige to grey to light brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text