Tributyrin, 97%
Tributyrin, 97%
Tributyrin, 97%
Thermo Scientific Chemicals

Tributyrin, 97%

Tributyrin, 97%, C15H26O6, CAS Number-60-01-5, butyrin, butyryl triglyceride, glycerol tributanoate, glycerol tributyrate, glyceroltributyrin, glyceryl tributyrate, propane-1,2,3-triyl tributyrate, tributin, tributyrin, tri-n-butyrin, 100mL, Liquid, 96% min. (GC), 02, 273, 305.0 deg.C to 310.0 deg.C | CAS: 60-01-5 | C15H26O6 | 302.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number 150881000
Price (USD)
67.65
Online Exclusive
75.00
Save 7.35 (10%)
Each
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
67.65
Online Exclusive
75.00
Save 7.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS60-01-5
IUPAC Name1,3-bis(butanoyloxy)propan-2-yl butanoate
Molecular FormulaC15H26O6
InChI KeyUYXTWWCETRIEDR-UHFFFAOYSA-N
SMILESCCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4330 to 1.4380 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
GC>=96.0 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0464521Certificate of AnalysisJul 18, 2024150882500, 150881000, 150880010
A0459380Certificate of AnalysisJan 16, 2024150882500, 150881000, 150880010
A0455276Certificate of AnalysisAug 25, 2023150882500, 150881000, 150880010
A0407134Certificate of AnalysisDec 13, 2022150882500, 150881000, 150880010
A0410170Certificate of AnalysisDec 12, 2022150882500, 150881000, 150880010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection