Thermo Scientific Chemicals

Tetraethylammonium chloride hydrate, 99%, Thermo Scientific Chemicals

Catalog number: 150905000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Tetraethylammonium chloride hydrate, 99%, Thermo Scientific Chemicals

Catalog number: 150905000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 150905000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
56-34-8
IUPAC Name
tetraethylazanium chloride
Molecular Formula
C8H20ClN
InChI Key
YMBCJWGVCUEGHA-UHFFFAOYSA-M
SMILES
[Cl-].CC[N+](CC)(CC)CC
Titration Argentometric
>=98.5 % (on dry substance)
Impurity
=<0.5 % Triethylamine hydrochloride
Appearance (Form)
Crystalline powder
Appearance (Color)
White
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text