L(-)-3-Phenyllactic acid, 98%, Thermo Scientific Chemicals
L(-)-3-Phenyllactic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

L(-)-3-Phenyllactic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 150920010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS20312-36-1
IUPAC Name(2S)-2-hydroxy-3-phenylpropanoate
Molecular FormulaC9H9O3
InChI KeyVOXXWSYKYCBWHO-QMMMGPOBSA-M
SMILESO[C@@H](CC1=CC=CC=C1)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or needles
TLC>=97.5 %
Infrared spectrumConforms
Appearance (Color)White to off-white
Melting point121°C to 125°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only