Thermo Scientific Chemicals

2-Amino-3-nitropyridine, 99%, Thermo Scientific Chemicals

Catalog number: 151120250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Amino-3-nitropyridine, 99%, Thermo Scientific Chemicals

Catalog number: 151120250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 151120250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4214-75-9
IUPAC Name
3-nitropyridin-2-amine
Molecular Formula
C5H5N3O2
InChI Key
BPYHGTCRXDWOIQ-UHFFFAOYSA-N
SMILES
NC1=NC=CC=C1[N+]([O-])=O
Infrared spectrum
Conforms
Melting point
164°C to 168°C
Appearance (Color)
Yellow
Appearance (Form)
Crystalline powder or needles
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text