1x1 image pixel for data collection
Pierce™ Streptavidin Coated Plates
Pierce™ Streptavidin Coated Plates
Pierce™ Streptavidin Coated Plates
Pierce™ Streptavidin Coated Plates
Thermo Scientific™
Pierce™ Streptavidin Coated Plates
Thermo Scientific™ Pierce Streptavidin Coated Plates are pre-blocked and ready-to-use polystyrene 96-well and 384-well plates for binding biotinylated antibodies or probes for ELISA and other target-specific assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorFormatPlate BlockingQuantity
15125ClearStandardBlocker™ BSA Buffer5 Plates
15219BlackStandardBlocker™ BSA Buffer5 Plates
15119BlackStandardSuperBlock™ Buffer5 Plates
15407BlackStandardSuperBlock™ Buffer5 Plates
15121ClearStrip PlateBlocker™ BSA Buffer5 Plates
15120ClearStrip PlateSuperBlock™ Buffer5 Plates
15122ClearStrip PlateSuperBlock™ Buffer5 x 5 Plates
15124ClearStandardSuperBlock™ Buffer5 Plates
15126ClearStandardSuperBlock™ Buffer5 x 5 Plates
15405ClearStandardSuperBlock™ Buffer5 Plates
15218WhiteStandardBlocker™ BSA Buffer5 Plates
15118WhiteStandardSuperBlock™ Buffer5 Plates
Catalog number 15125
Price (USD)
238.65
Online Exclusive
265.00
Save 26.35 (10%)
Each
Add to cart
Color:
Clear
Format:
Standard
Plate Blocking:
Blocker™ BSA Buffer
Quantity:
5 Plates
Request bulk or custom format
Price (USD)
238.65
Online Exclusive
265.00
Save 26.35 (10%)
Each
Add to cart
Streptavidin-coated microplates provide the biotin-binding function needed for a variety of ELISA and other assay procedures based on biotinylated capture antibodies, peptide antigens, and other analytes such as oligonucleotides. Pierce Streptavidin Coated Plates are available in clear, white, and black plates for use with colorimetric, chemiluminescent, and fluorescent detection methods, respectively.
Thermo Scientific Pierce Streptavidin Coated Plates are pre-blocked and ready-to-use polystyrene plates for binding biotinylated antibodies or probes for ELISA and other target-specific assays.
Features of Streptavidin Coated Plates include:
• Approximate binding capacity is ∼5 pmol biotin/well
• Easy and gentle immobilization of biotin-containing conjugates
• Low nonspecific binding
• No denaturing of the protein component of a conjugate upon binding
• Ideal for binding small biotinylated hydrophilic molecules (e.g., peptides) that typically exhibit poor binding to polystyrene
• Pre-blocked with your choice of Blocker BSA or SuperBlock Blocking Buffer
Biotin-binding plates are ideal for applications when direct adsorption to polystyrene denatures antibodies or the target molecule. Streptavidin is the most popular and widely available biotin-binding protein for research methods. Streptavidin has an isoelectric point of 5 to 6, resulting in low nonspecific interactions.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Binding PropertyStandard Capacity, Affinity
Detection MethodColorimetric
Plate BlockingBlocker™ BSA Buffer
Product TypeDetection Plate, Immunoassay, ELISA
Quantity5 Plates
ColorClear
FormatStandard
MaterialPolystyrene
No. of Wells96
Product LinePierce
Surface TreatmentStreptavidin
Unit SizeEach
Contents & Storage
Upon receipt store plates at 4°C in unopened pouches. Once opened, place unused plates in a resealable bag with desiccant and store at 4°C. Plates are shipped at ambient temperature.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link