Thermo Scientific Chemicals

Chlorotriphenylsilane, 95%, Thermo Scientific Chemicals

Catalog number: 151251000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Chlorotriphenylsilane, 95%, Thermo Scientific Chemicals

Catalog number: 151251000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 151251000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
76-86-8
IUPAC Name
chlorotriphenylsilane
Molecular Formula
C18H15ClSi
InChI Key
MNKYQPOFRKPUAE-UHFFFAOYSA-N
SMILES
Cl[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to almost white
Appearance (Form)
Powder or crystalline powder or
Appearance (Form)
crystals or chunks
Infrared spectrum
Conforms
Melting point
91°C to 97°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text