Acetyl chloride, 98%, Thermo Scientific Chemicals
Acetyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 151270010
Price (USD)/ Each
94.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
111.00 
Save 16.35 (15%)
-
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
94.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
111.00 
Save 16.35 (15%)
Add to cart
Acetyl chloride, 98%, Thermo Scientific Chemicals
Catalog number151270010
Price (USD)/ Each
94.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
111.00 
Save 16.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameacetyl chloride
Molecular FormulaC2H3ClO
InChI KeyWETWJCDKMRHUPV-UHFFFAOYSA-N
SMILESCC(Cl)=O
Molecular Weight (g/mol)78.50
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.3876 to 1.3896 (20°C, 589 nm)
Color scale=<30 APHA
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only