Bromocresol Green, pure, indicator grade
Bromocresol Green, pure, indicator grade
Bromocresol Green, pure, indicator grade
Bromocresol Green, pure, indicator grade
Thermo Scientific Chemicals

Bromocresol Green, pure, indicator grade

Bromocresol Green, C21H14Br4O5S, CAS Number-76-60-8, 3', 3″, 5', 3',3″,5',5″-Tetrabromo-m-cresolsulfonephthalein, BCG, 5g, Brown, 200-972-8, 698.01, Pure, Warning, GHS H Statement: May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation. | CAS: 76-60-8 | C21H14Br4O5S | 698.014 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1513100505 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 151310050
Price (USD)
53.65
Online Exclusive
59.90
Save 6.25 (10%)
Each
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
53.65
Online Exclusive
59.90
Save 6.25 (10%)
Each
Add to cart
Chemical Identifiers
CAS76-60-8
IUPAC Name3,3-bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular FormulaC21H14Br4O5S
InChI KeyFRPHFZCDPYBUAU-UHFFFAOYSA-N
SMILESCC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C(O)C(Br)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
UV Lambda max613 to 621 nm (in buffer)
Visual transition intervalfrom pH 3.8 (yellow) to pH 5.4 (blue)
Appearance (Color)Yellow to beige to light brown
Solubility(0.1 % in ethanol) Clear
Heavy metals (as Pb)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Share catalog number, name or link