Thermo Scientific Chemicals

Bromophenol Blue, sodium salt, pure, water soluble, indicator

Catalog number: 151350250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Bromophenol Blue, sodium salt, pure, water soluble, indicator

Catalog number: 151350250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 151350250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
34725-61-6
IUPAC Name
sodium 3,5-dibromo-4-[3-(2,6-dibromo-4-hydroxyphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]benzen-1-olate
Molecular Formula
C19H9Br4NaO5S
InChI Key
KGANHHDZDYZJEM-UHFFFAOYNA-M
SMILES
[Na+].OC1=CC(Br)=C(C(Br)=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=C(Br)C=C([O-])C=C1Br
UV spectrum
Complies
Visual transition interval
from pH 3.0 (yellow) to pH 4.6 (blue)
Infrared spectrum
Conforms
Clarity of solution
(0.1% in water) clear
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text